missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INTS10, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant INTS10.
Brand: Abnova PAB23514.100uL
This item is not returnable.
View return policy
Description
INTS10 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1; MIM 180680) and U2 (RNU2; MIM 180690) (Baillat et al., 2005 [PubMed 16239144]).[supplied by OMIM
Sequence: KGRRSYGDILHRMKDLCRYMNNFDSEAHAKYKNQVVYSTMLVFFKNAFQYVNSIQPSLFQGPNAPSQVPLVLLEDVSNVYGDSpecifications
INTS10 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant INTS10. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
C8orf35/FLJ10569/INT10 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
55174 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry, Immunohistochemistry (Formalin/Paraffin) | |
Unconjugated | |
Immunohistochemistry (1:1000-1:2500) The optimal working dilution should be determined by the end user. | |
INTS10 | |
INTS10 | |
Recombinant protein corresponding to amino acids of human INTS10. | |
100 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction