Learn More
LRBA Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LRBA.
Brand: Abnova PAB21205.100uL
Description
Patients with Chediak-Higashi syndrome (CHS1; MIM 214500) suffer from a systemic immunodeficiency involving defects in polarized trafficking of vesicles in a number of immune system cell types. In mouse, this syndrome is reproduced in strains with a mutation in the 'beige' gene that results in proteins lacking the BEACH (beige and CHS1) domain and C-terminal WD repeats. LRBA contains key features of both beige/CHS1 and A kinase anchor proteins (AKAPs; see MIM 602449).[supplied by OMIM
Sequence: SVLMVSKYRDILEPQNERHSQSCTETGSENENVSLSEITPAAFSTLTTASVEESESTSSARRRDSGIGEETATGLGSHVEVTPHTAPPGVSAGPDAISEVLSTLSLEVNKSPETKNDRGNDLDTKATPSVSVSpecifications
LRBA | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant LRBA. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
BGL/CDC4L/DKFZp686A09128/DKFZp686K03100/DKFZp686P2258/FLJ16600/FLJ25686/LAB300/LBA/MGC72098 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
987 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
LRBA | |
LRBA | |
Recombinant protein corresponding to amino acids of human LRBA. | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only