Learn More
Rabbit anti-Monkey Serum Proteins Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69804
Product Code. 18613719
  • £209.00 / 1mL
Explore more special offers
Description
Description
Serum Proteins Polyclonal antibody specifically detects Serum Proteins in Monkey samples. It is validated for ELISA, Immunoprecipitation, Immunoelectrophoresis
Specifications
Specifications
Serum Proteins
Polyclonal
Unconjugated
Whole antiserum
Pooled whole Rhesus monkey (Macaca mulatta) serum and partly purified serum fractions. Freund's complete adjuvant is used in the first step of the immunization procedure.
1 mL
Secondary
Monkey
Lyophilized
ELISA, Immunoprecipitation, Immunoelectrophoresis
LYOPH
ELISA, Immunoprecipitation 1:10 - 1:500, Immunoelectrophoresis
Rabbit
Whole Antiserum
RUO
Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2).
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.