missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit IgG Isotype Control, BD
Brand: BD Biosciences 550875
Additional Details : Weight : 0.01000kg
Description
Host Species: Rabbit
Isotype: IgG
Applications:
ELISA Standard (Routinely Tested)
Immunohistochemistry, Flow cytometry (Not Recommended)
Specifications
ELISA | |
Unconjugated | |
Trinitrophenol-KLH | |
0.5 mg | |
IgG |
0.5mg/mL | |
Rabbit | |
Affinity Purified | |
Undiluted at 4°C |