missing translation for 'onlineSavingsMsg'
Learn More

Rabbit IgG Isotype Control, Invitrogen™

Rabbit Isotype Control

Brand:  Invitrogen 10500C

Code : 50

Additional Details : Weight : 0.50000kg

Product Code. 10312573

  • £108.00 / Each
Estimated Shipment: 26-07-2024
to see stock.
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

Rabbit IgG refers to immunoglobulin G antibodies produced by rabbits in response to an antigen. These antibodies are commonly used in laboratory research as a detection tool for specific proteins, molecules, or cells. Rabbit IgG antibodies are highly specific and have a strong affinity for their target, making them ideal for use in various applications such as western blotting, immunohistochemistry, and flow cytometry.
Specifications

Specifications

Rabbit IgG
3 mg/mL
PBS with 0.1% sodium azide
Rabbit
purified
RUO
Not Applicable
Liquid
Control, Flow Cytometry
Unconjugated
Immunoglobulin G; ImmunoglobulinG; Not Applicable
Normal Rabbit IgG
5 mL
Isotype Control (Primary)
4° C
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers