missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rabbit IL-2 ELISA Kit
Rabbit IL-2 quantitates rabbit IL-2 in serum, plasma, supernatant.
£530.00 - £2320.00
Specifications
Accession Number | Aldesleukin; cytokine; H-IL-2; il 2; Il2; IL-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; Interleukin2; interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; TCGF |
---|---|
Assay Range | 2.4 to 600 pg/mL |
Assay Sensitivity | 2.4 pg/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17185839
|
Invitrogen™
ER7RBX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
17105023
|
Invitrogen™
ER7RB |
96 Tests |
£530.00
Pack of 96 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
Description
Rabbit IL-2 quantitates rabbit IL-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rabbit IL-2.
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Specifications
Aldesleukin; cytokine; H-IL-2; il 2; Il2; IL-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; Interleukin2; interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; TCGF | |
2.4 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
IL2 | |
<10% | |
HRP | |
Plasma, 50 μL; Supernatant, 50 μL; Serum, 50 μL | |
IL-2 | |
4 hr 45 min |
2.4 to 600 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
O77620 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |