missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Rabbit Serum

This rabbit serum is derived from the blood of healthy adult animals. Rabbit serum is used to support cultures of Borrelia burgdorferi, the causative agent of Lyme disease.

Brand:  MP Biomedicals™ 092941154

Additional Details : Weight : 0.50000kg

Product Code. 17388573

  • £242.74 / 500mL
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

This rabbit serum is derived from the blood of healthy adult animals and is used to support cultures of Borrelia burgdorferi, the causative agent of Lyme disease.

Usage Statement: Unless specified otherwise, MP Biomedical™ products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Specifications

Specifications

Adult
Liquid
500 mL
Sterile
Support Cultures of Borrelia burgdorferi
Rabbit
Rabbit Serum
4 to 6.6 g/dL
0.2 μm filtration
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers