missing translation for 'onlineSavingsMsg'
Learn More

RAC2 Mouse anti-Human, Mouse, Clone: 4C2H6, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60077-1-IG-20UL

 View more versions of this product

Product Code. 16857045

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

RAC, a small GTPase in the ras family, regulates the formation of membrane ruffles, lamellipodia and filopodia. It is involved in cytoskeletal actin organization and transformation. The p21 activated kinases (PAKs) are direct targets of active Rac and Cdc42 which can induce the assembly of polarized cytoskeletal structures when expressed in fibroblasts. RAC1 and RAC2 are also essential components of NADPH oxidase, the enzyme responsible for generating free radicals.
Specifications

Specifications

RAC2
Monoclonal
1.5 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P15153, Q05144
RAC2
RAC2 Fusion Protein Ag1202
20 μL
Primary
Mouse, Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
4C2H6
Unconjugated
RAC2
EN 7, Gx, HSPC022, p21 Rac2, RAC2, Small G protein
Mouse
Protein G
RUO
19354, 5880
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers