Learn More
Fisherbrand™ Rack for Glassware, 3 Levels
The rack is constructed of stainless steel and fitted with a top load indicator.
Brand:  Fisherbrand™
Additional Details : Weight : 1.04000kg
Product Code. 17167794
  • £1764.90 / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
  • Top load indicator ensures that the glassware is not damaged when being pushed into the washing chamber
Specifications
Specifications
3 Levels
Stainless Steel
Rack
Glassware