Learn More
Thermo Scientific™ Specialty Racks
Maximize the storage space in your ultra-low temperature freezer with these racks.
Brand:  Thermo Scientific™ 920022
 View more versions of this product
Product Code. 10022573
  • £207.66 / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Microplate racks in standard or deepwell formats
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Side Access Freezer Rack (Adjustable)
Silver
Stainless Steel
5
13.5 cm
29.5 cm
24.6 in.
2 in. Box Racks - Adjustable side access rack
Boxes (2 in.)
22 Boxes/Rack
17.3 and 23 cu. ft. capacity upright freezers
5.3 in.
11.6 in.
62.5 cm