Learn More
Thermo Scientific™ Specialty Racks
Maximize the storage space in your ultra-low temperature freezer with these racks.
Brand:  Thermo Scientific™ 398322
 View more versions of this product
Product Code. 11379311
  • £196.64 / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • Warranty varies by country, please contact us for details
  • TRUSTED_SUSTAINABILITY
Specifications
Specifications
Side Access Freezer Rack (Microplate/Locking Rod)
Silver
Stainless Steel
Rectangular
Revco 32 cuft Upright ULT
5.5 in.
9.4 in.
65.3 cm
Side Access Microplate Rack w/Locking Rod - Standard or Deepwell
Microplates or Deepwell Microplates
98 microplates with lids, 119 microplates without lids, and 35 deepwell
4 Tiers
14 cm
23.9 cm
25.7 in.