Learn More
Thermo Scientific™ Specialty Racks
Maximize the storage space in your ultra-low temperature freezer with these racks.
Brand:  Thermo Scientific™ 1950651
 View more versions of this product
Product Code. 12662725
  • £228.85 / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Microplate racks in standard or deepwell formats
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Side Access Freezer Rack (Microplate/Deepwell)
Silver
Stainless Steel
Square
14.0 cm
30.2 cm
14.7 in.
Side Access Microplate Rack - Standard or Deepwell
Microplates
56 Plates/Rack
13 cu. ft. capacity upright freezers
5.5 in.
11.9 in.
37.3 cm