Learn More
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Brand:  Thermo Scientific™ 1600084
 View more versions of this product
Product Code. 11927284
  • £156.61 / Pack of 2
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rack Insert; Includes top and bottom panels
Rack Insert
Stainless Steel
Bath Types S13, S12T, S15
39 10 mm Tubes