Learn More
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer.
Brand:  Thermo Scientific™ 4000701
 View more versions of this product
Product Code. 10633575
  • £383.12 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
  • Warranty varies by country, please contact us for details
  • TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black
All CryoMed controlled-rate freezers
Vial Rack (Small)
76 1.2 mL/2 mL Ampules
76 1.2 mL/2 mL Ampules
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer > Small rack for 1.2 and 2mL vials (holds 76)