Learn More
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer.
Brand:  Thermo Scientific™ 4000303
 View more versions of this product
Product Code. 11726499
  • £740.80 / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
  • Warranty varies by country, please contact us for details
  • TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black
CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458
Cane Freezing Rack
162 Canes, 5 to 6 Ampules/Cane
162 Canes, 5 to 6 Ampules/Cane
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer > Cane freezing rack (162 canes)