missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD51 Monoclonal Antibody (13E4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA123330
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa whole cell extract, recombinant human Rad51 protein.
RAD51 plays a critical role in homologous strand exchange, a key step in DNA repair through homologous recobination. It binds to single and double-stranded DNA and exhibits DNA-dependent ATPase activity. RAD51 catalyzes the recognition of homology and strand exchange between homologous DNA partners to form a joint molecule between a processed DNA break and the repair template. It binds to a single-stranded DNA in an ATP-dependent manner to form nucleoprotein filaments which are essential for the homology search and strand exchange. Mutations in the gene can results in breast cancer, mirror movements 2 and fanconi anemia.Specifications
RAD51 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
Q06609, Q08297 | |
Rad51 | |
Recombinant human Rad51 protein. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Western Blot | |
13E4 | |
Unconjugated | |
Rad51 | |
AV304093; BRCA1/BRCA2-containing complex, subunit 5; BRCC5; DNA repair protein RAD51; DNA repair protein RAD51 homolog 1; DNA repair protein RAD51 homolog A; DNA repair protein rhp51; FANCR; homolog to S.cerevisiae; HRAD51; HsRad51; HsT16930; MGC128559 protein; MRMV2; MUT5; rad51; RAD51 homolog; RAD51 homolog (RecA homolog); RAD51 homolog (RecA homolog, E. coli); RAD51 homolog A; RAD51 recombinase; RAD51 recombinase L homeolog; rad51.1; rad51.L; rad51-1; RAD51A; rad51-a; rad51-b; Reca; RecA family recombinase Rhp51; RecA, E. coli, homolog of; RecA-like protein; recombinase RAD51; recombination protein A; RGD1563603; rhp51; SPAC644.14c; wu:fb38e12; xrad51; XRad51.1; YER095W; zgc:77754 | |
Mouse | |
Protein G | |
RUO | |
19361, 5888 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |