Learn More
Thermo Scientific™ RadEye™ PRD/PRD-ER Personal Radiation Detector
Locate hidden radiation sources generated by nuclear weapons, improvised nuclear devices or radiological dispersal devices with high-performance radiation detection technology.
Brand:  Thermo Scientific™ 425067122
 View more versions of this product
Product Code. 10470285
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Show More
Specifications
Specifications
RadEye PRD-ER, Factory Calibrated in H*(10) μSv/h
Personal Radiation Detector
NaI(Tl)-detector with high-quality microphotomultiplier
For Cs-137 (662 kev): 1.5 cps per µR/hFor Am-241 (60 kev): 30 cps per µR/hFor Cs-137 (662 kev): 150 cps per µSv/hFor Am-241 (60 kev): 2000 cps per µSv/h
Gamma
60 keV to 1.3 MeV
1 μrem/h to 10 rem/h (0.01 μSv/h to 100 mSv/h)
10000 rem/h (100 Sv/h)
Interdiction and response
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ RadEye™ PRD/PRD-ER Personal Radiation Detector > Factory calibrated in H(10) microSieverts/hr.; Yellow