Learn More
Fisherbrand™ Radiation Safety Gamma Boxes
This gamma radiation bin is ideal for holding microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes.
Brand:  Fisherbrand™ CSR-B2TIPG
Product Code. 12357117
  • £2070.00 / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  •  Also available is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids, which serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, and the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
Specifications
Specifications
CSR-BAG1
13mm x 13mm x 13mm
2 L
130mm
130mm