Learn More
Cytiva Rainbow™ Molecular Weight Markers
Monitor progress of protein electrophoresis and assess transfer efficiency and molecular weight of blotted proteins without staining. Cytiva Life Sciences™ Rainbow™ Molecular Weight Markers use bright, distinctive colors to allow easier confirmation of transfer to blotting membranes and orientation.
Brand: Cytiva RPN800E
203.91 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : GE
Additional Details : CAS Number : 56-81-5 Weight : 0.01350kg
Description
Enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels. The ready-to-load markers provide sharper, more intense bands on gels and blots and have improved band spacing for more accurate molecular weight determination.
- High intensity, prestained multicolored bands allow distinction of closely related proteins
- Full-Range Rainbow Markers (Mr 12,000 to 225,000) use ten separate proteins with six different colors
- High-Range Rainbow Markers (Mr 12,000 to 225,000) use eight separate proteins with six different colors
- Low-Range Rainbow Markers (Mr 3500 to 38,000) use seven separate proteins with five different colors
- For use on SDS-PAGE
- The bright, distinctive colors of the markers allow easier confirmation of transfer to blotting membranes and orientation.
- Markers are a mixture of individually colored proteins that are combined to produce bands of equal color intensity. Approximate protein concentration is 1 to 2 mg/mL, depending on the product
- Each 250μL pack size is sufficient for use with 50 minigels (10 × 8cm) or 25 large gels (20 × 20cm)
Specifications
Rainbow molecular weight markers enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels. | |
2 mg/mL | |
Liquid | |
For industrial applications: Analytical reagent, research Use ten separate proteins with six different colors | |
250 μL | |
-15 to -30°C |
Multi | |
Protein Molecular Weight Marker/Ladder | |
Supplied ready to use in 305% Glycerol and sample Buffer containing, Mercaptoethanesulphonic Acid (MESNA) as reducing agent | |
Mr 12 000 | |
12000 to 225000Mr | |
50 Minigels or 25 large gels |
Safety and Handling
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash hands thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : sodium dodecyl sulphate, mesna, Glycine,N, N'-1, 2-ethanediylbis¢N-(carboxymethyl)-, sodium salt, h
- UNKNOWN ACUTE-35.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 32.5% of components with unknown hazards to the aquatic environment.