Learn More
Mettler Toledo™ Rainin™ BioClean™ Bulk Pipette Tips
Product Code. p-8006186
Encompass_Preferred
Click to view available options
Volume (Metric):
20 μL
1000 μL
Unit Size:
Pack of 1000
Pack of 200
Product Code. 12089001
Brand: Mettler Toledo™ 17001128
Only null left
Add to basket
Add to basket
BioClean™ tips provided in economical bulk bags for use with Rainin™ pipettes with LTS™ LiteTouch™.
  • Flawless performance: perfect production and advanced design
  • No deformations, cracks or flashes
  • 100% inert: purity and cleanliness are assured
  • All tips are 100% free of DNA, DNAse, RNAse, ATP, Pyrogens, PCR inhibitors and additives
  • Packaged in a re-sealable zip-lock bag, for manual rack refilling
Specifications
Volume (Metric) 20 μL
Filtered Non-filtered
Sterility Non-sterile
Packaging 1000 Tips/Bag
For Use With (Equipment) Rainin LTS LiteTouch
Product Line Rainin BioClean