missing translation for 'onlineSavingsMsg'
Learn More

Raloxifene hydrochloride, Tocris Bioscience™

Product Code. 18752584
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective estrogen receptor modulator (SERM)

Selective estrogen receptor modulator (SERM) that binds to ERα and ERβ, and tissue-dependently activates or blocks estrogen-induced transcription. Acts as an antiestrogen in breast and uterine tissue, but displays estrogen agonist activity in bone. In D12 rat hypothalamic cells, inhibits progesterone receptor induction by estrogen with an IC50 of 1 nM.

Specifications

CAS 82640-04-8
Molecular Formula C28H28ClNO4S
Quantity 50 mg
Synonym raloxifene hydrochloride, raloxifene hcl, evista, keoxifene hydrochloride, 6-hydroxy-2-4-hydroxyphenyl benzo b thiophen-3-yl 4-2-piperidin-1-yl ethoxy phenyl methanone hydrochloride, keoxifene, unii-4f86w47br6, evista raloxifene hydrochloride, 6-hydroxy-2-p-hydroxyphenyl benzo b thien-3-yl-p-2-piperidinoethoxy phenyl ketone, hydrochloride
InChI Key BKXVVCILCIUCLG-UHFFFAOYSA-N
SMILES C1CCN(CC1)CCOC2=CC=C(C=C2)C(=O)C3=C(SC4=C3C=CC(=C4)O)C5=CC=C(C=C5)O.Cl
IUPAC Name [6-hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[4-(2-piperidin-1-ylethoxy)phenyl]methanone;hydrochloride
Molecular Weight (g/mol) 510.045
PubChem CID 54900
ChEBI CHEBI:50740
Formula Weight 510.04
Percent Purity >98%
Chemical Name or Material Raloxifene hydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.