missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALYL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541957
This item is not returnable.
View return policy
Description
Peptide sequence: AQKKQLEESL VLIQEECVSE IADHSTEEPA EGGPDADGEE MTDGIEEDFD Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 79%; Pig: 100%; Rabbit: 93%
RALYL Polyclonal antibody specifically detects RALYL in Human samples. It is validated for Western Blot.Specifications
RALYL | |
Polyclonal | |
Unconjugated | |
RALYL | |
0710005M24Rik; Heterogeneous nuclear ribonucleoprotein C-like 3; hnRNP core protein C-like 3; HNRPCL3; hRALYL; RALY RNA binding protein like; RALY RNA binding protein-like; RALYL; RNA-binding Raly-like protein; Unknown (protein for MGC:133552) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
138046 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86SE5 | |
RALYL | |
Synthetic peptide directed towards the C-terminal of human RALYL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |