missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Raney™-Nickel, Activated Catalyst, 50% slurry in water, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 7440-02-0, 7732-18-5, 7429-90-5 |
---|---|
Molecular Formula | Ni |
Molecular Weight (g/mol) | 58.69 |
MDL Number | MFCD00011137 MFCD06798735 |
InChI Key | PXHVJJICTQNCMI-UHFFFAOYSA-N |
Synonym | raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum |
PubChem CID | 935 |
ChEBI | CHEBI:28112 |
IUPAC Name | nickel |
SMILES | [Ni] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-02-0, 7732-18-5, 7429-90-5 | |
58.69 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
935 | |
nickel |
Ni | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
CHEBI:28112 | |
[Ni] |
Specifications
7440-02-0, 7732-18-5, 7429-90-5 | |
MFCD00011137 MFCD06798735 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 | |
58.69 | |
Slurry |
Ni | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
[Ni] | |
58.69 | |
CHEBI:28112 | |
Plastic bottle | |
Raney™-Nickel, Activated Catalyst |
Safety and Handling
missing translation for 'einecsNumber' : 231-111-4
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Raney Nickel, activated catalyst, 50% slurry in water
Signal Word
- Danger
Hazard Category
- Self-heating substance Category 1
- Specific target organ toxicity Category 1
- Skin sensitiser Category 1
- Carcinogenicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H251-Self-heating: may catch fire.
- H372-Causes damage to organs.
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P235+P410-Keep cool. Protect from sunlight.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
Supplemental information
- MIXTURE LIST-Contains : Nickel, Aluminium powder
RUO – Research Use Only