Learn More
RANGAP1 Mouse anti-Human, Unconjugated, Clone: 1F3A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67146-1-IG-20UL
This item is not returnable.
View return policy
Description
The RANGAP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RANGAP1. This antibody recognizes human, mouse, rat antigen. The RANGAP1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
RANGAP1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunofluorescence, ELISA | |
1F3A4 | |
Unconjugated | |
20 μL | |
5905 | |
IgG1 |