Learn More
OHAUS™ Ranger™ 7000
OHAUS™ Ranger™ 7000 boasts a high resolution, has a large display, a variety of connectivity options, and a large memory.
£1315.00
Specifications
Connection Type Easy access communication port including standard USB and RS232 (included) and an optional third port of RS232 or Ethernet (accessories sold separately). GLP/GMP Data Output with Real Time Clock. Data Transfer Function.
Construction Modular design, die-cast housing, removable stainless steel platform, integral weigh below hook, menu lock switch, level indicator
Display Type Full-color WQVGA Graphic Display with user-controlled brightness
Protection Class IP54
Weighing Modes Gram, Kilogram, Ounce, Pound, Pound:Ounce, Custom
View More Specs
Products 1
Product Code Brand Capacity Price Quantity & Availability  
Product Code Brand Capacity Price Quantity & Availability  
15614553
View Documents Promotion Details
OHAUS™
30088847
35.00 kg
£1315.00
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Description
Description
  • Simplify complex applications and minimize the need for manual calculations with 10 advanced application modes, peripheral device control and an optional kit for a second scale platform.
  • The most precise scale in its class with one-second stabilization time, up to 75,000d or 350,000d display resolution and legal-for-trade certifications.
  • Extremely durable scale designed to thrive in rugged industrial environments.
  • Features IP54-rated metal housing and sealed metal terminal.
Specifications
Specifications
Easy access communication port including standard USB and RS232 (included) and an optional third port of RS232 or Ethernet (accessories sold separately). GLP/GMP Data Output with Real Time Clock. Data Transfer Function.
Full-color WQVGA Graphic Display with user-controlled brightness
Gram, Kilogram, Ounce, Pound, Pound:Ounce, Custom
Modular design, die-cast housing, removable stainless steel platform, integral weigh below hook, menu lock switch, level indicator
IP54