Learn More
Ohaus™ Ranger™ 7000 Bench Scale
Features high resolution, large display, many application modes and connectivity options, large memory.
Brand:  Ohaus™ 30088850
Product Code. 15802942
  • £1450.00 / Each
Explore more special offers
Description
Description
  • Advanced features for next-level industrial weighing
  • Fast performance and legal for trade certifications assure accurate results
  • Sturdy industrial and modular design support flexible and heavy-duty use
  • Cutting edge SmarText™ software
  • Standard connectivity with flexible options
  • Print templates
TRUSTED_SUSTAINABILITY
Specifications
Specifications
12 Hours with Rechargeable Battery
USB (Included), RS232 (Included), 2nd RS232 (Available as an Accessory), Ethernet (Available as an Accessory)
15 kg
0.200000 g
0.400000 g
R71MD15-GB
467.00 x 377.00 x 128.000 mm
467.00 mm
14.90 in.
377 mm
128.000 mm
467.00 mm
Not Applicable
Modular design, die-cast housing, removable stainless steel platform, integral weigh below hook, menu lock switch, level indicator
TFT Graphic LCD, Full-color WQVGA Graphic Display with user-controlled brightness
USB, RS232, in-use cover, AC power adapter (rechargeable battery not included)
Stainless Steel Platform
467.00 x 377.00 mm
18.40 in.
311 mm
377.00 mm
5.00 in.
18.40 in.
100/240 VAC 50/60 Hz Universal Power Supply, rechargeable lithium battery (Optional)