Description
- Advanced features for next-level industrial weighing
- Fast performance and legal for trade certifications assure accurate results
- Sturdy industrial and modular design support flexible and heavy-duty use
- Cutting edge SmarText™ software
- Standard connectivity with flexible options
- Print templates
Specifications
Specifications
| Battery Life | 12 Hours with Rechargeable Battery |
| Calibration | Not Applicable |
| Connection Type | USB (Included), RS232 (Included), 2nd RS232 (Available as an Accessory), Ethernet (Available as an Accessory) |
| Construction | Modular design, die-cast housing, removable stainless steel platform, integral weigh below hook, menu lock switch, level indicator |
| Capacity | 15 kg |
| Display Type | TFT Graphic LCD, Full-color WQVGA Graphic Display with user-controlled brightness |
| Readability | 0.200000 g |
| Includes | USB, RS232, in-use cover, AC power adapter (rechargeable battery not included) |
| Linearity | 0.400000 g |
| Material | Stainless Steel Platform |
| Show More |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering