Learn More
Ohaus™ Ranger™ Count 4000 Counting Scale
Suitable for tough, industrial counting jobs that require durability and accuracy at a great value.
Marke: Ohaus™ 30231105
798.49 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Equipped with weighing, parts counting, check counting, and accumulation functions, and can produce results for a variety of industrial weighing, counting, and measurement needs
- Full metal housing offers strength and protection and maximizes product life
- Precise counting and weighing performance produce accurate results
- User-friendly software
- Easy to operate
- Several connectivity options provide connection to multiple peripheral devices
Spezifikation
210 Hours with Rechargeable Battery | |
Rugged Cast Aluminum Housing, Stainless Steel Platform | |
3-window Backlight LCD + 3 LED | |
RS232, AC Power, Rechargeable Battery | |
Rugged Cast Aluminum Housing, Stainless Steel Platform | |
2.000000 g | |
327.00 x 315.00 x 122.000 mm | |
327.00 mm | |
12.40 in. | |
300 mm | |
122.000 mm | |
327.00 mm |
2nd RS232 (Available as an Accessory)Ethernet (Available as an Accessory)RS232 (Included)USB (Available as an Accessory) | |
30 kg | |
1.000000 g | |
2.000000 g | |
RC41M30 | |
327.00 x 315.00 mm | |
12.90 in. | |
225 mm | |
315.00 mm | |
4.80 in. | |
12.90 in. | |
100/240 VAC 50/60 Hz internal universal power supply, internal rechargeable lead acid battery |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur