Learn More
Novex™ Rapamycin
Rapamycin
Brand: Novex™ PHZ1235
262.67 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 53123-88-9 Weight : 0.01000kg
Description
Rapamycin is a member of a family of macrolide immunosuppressants that bind to and inhibit the FK506 binding protein (FKBP) proline rotamase.Format/Formulation: Lyophilized
Specifications
Store in freezer (-5 to -30 °C). | |
MTOR | |
mechanistic target of rapamycin (serine/threonine kinase) | |
Rapamycin | |
Solid |
2475 | |
FRAP, FRAP1, FRAP2, RAFT1, RAPT1 | |
Wet Ice | |
914.2 |
Safety and Handling
- RAPAMYCIN
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see supplemental first aid instructions on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Rapamycin
For Research Use Only. Not for use in diagnostic procedures.