missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rapamycin
Click to view available options
Quantity:
1 mg
Description
Rapamycin is a antifungal and immunosuppressant. Specific inhibitor of mTOR (mammalian target of Rapamycin). Complexes with FKBP-12 and binds mTOR inhibiting its activity. Inhibits interleukin-2-induced phosphorylation and activation of p70 S6 kinase. Induces autophagy in yeast and mammalian cell lines. Drives hPSC differentiation to mesendoderm and blood progenitor cells. Also used as a chemical dimerizer; rapamycin and GA3-AM (Cat. No. 5407) chemically inducible dimerization systems are orthogonal. Inhibits MERS-CoV infection of Huh7 cells in a plaque reduction assay. Enhances lentiviral transduction of hematopoietic stem cells.
Specifications
Type | Rapamycin |
CAS | 53123-88-9 |
Molecular Formula | C51H79NO13 |
Synonym | (3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epepoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone |
InChI Key | QFJCIRLUMZQUOT-HPLJOQBZSA-N |
SMILES | CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)O)C)C)O)OC)C)C)C)OC |
Molecular Weight (g/mol) | 914.187 |
PubChem CID | 5284616 |
ChEBI | CHEBI:9168 |
Formula Weight | 914.18 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'