missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapID™ ANA II System

Identify over 90 medically important anaerobes in just four hours using this rapid identifcation system based on enzyme technology.

Brand:  Thermo Scientific™ R8311002

254.08 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Product Code. 11464139

  • £410.00 / Pack of 20

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

2°C to 8°C
Yes
1 Box
RapID ANA II System
Anaerobes
20 Panels/Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360Df-May damage the unborn child. Suspected of damaging fertility.

Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ RapID™ ANA II System > 20 Panels/Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.