Learn More
3M™ Petrifilm™ Rapid Coliform Count Plate
Designed to provide fast, accurate results in detecting coliform contamination.
£90.95 - £836.70
Specifications
Test Type Environmental Test
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17203754
Encompass
View Documents
3M™ Petrifilm™
7100039311
50
£90.95
Pack of 50
View alternative products
Save Up to 
»
Estimated Shipment: 14-02-2025
to see stock.
Add to basket
17233754
Encompass
View Documents
3M™ Petrifilm™
7100039395
500
£836.70
Pack of 500
View alternative products
Save Up to 
»
Estimated Shipment: 14-02-2025
to see stock.
Add to basket
Description
Description
  • Utilizes the convenience and efficiency of the 3M™ Petrifilm™ Plate platform.
  • Proven cost effective tool for the detection of coliform.
  • Results in 4-24 hours.
  • This plate has been awarded Official Methods of Analysis (OMA) #2000.15 for Food and NF VALIDATION certificates: 3M-01/5-03/97 A for all human food products , 3M-01/5-03/97 B for All human food products and 3M-01/5-03/97 C for All human food products (except process pork products) by AFNOR Certification.
  • Food processors and contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Specifications
Environmental Test