Learn More
Thermo Scientific™ Pierce™ Rapid Gold BCA Protein Assay Kit
Thermo Scientific™ Pierce™ Rapid Gold BCA Protein Assay Kit is two-component, high-precision, detergent-compatible assay optimized to measure (at 480 nm) total protein concentration compared to protein standard curve of known concentrations.
Brand: Thermo Scientific™ A53225
Alert:
Store at 4°C
Description
- A fast, two-component, high-precision, detergent-compatible assay optimized to determine total protein concentration
- Can be used to assess yields in whole cell lysates and affinity-column fractions, as well as to monitor protein contamination in industrial applications
- Assay maintains the key characteristics of the traditional BCA assay but allows a fast time and room temperature incubation equal to dye-binding methods
- Provides comparable accuracy but with a 5 min., room temperature incubation and measured at 480 nm
- This improvement eliminates the need to wait or expose the samples to elevated temperatures for a fast time to results
- Exhibits similar protein-to-protein variation as traditional BCA Protein Assay and less protein-to-protein variation than dye-binding protein assay methods (Bradford)
- Unaffected by typical concentrations of most ionic and non-ionic detergents
- Linear working range for BSA of 20 to 2000 μg/mL
- Store in refrigerator at 2°C to 8°C
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
20 to 2000 μg/mL | |
Rapid Gold BCA Assay | |
Pierce™ | |
Not Target-Specific | |
Spectrophotometer, Microplate Reader | |
Wet Ice |
Store in refrigerator (2–8°C). | |
Colorimetric | |
Protein Quantitation Assay | |
Solution-based Detection, Absorbance | |
500 mL |
Safety and Handling
- Pierce Rapid Gold BCA Protein Assay Kit
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P264-Wash hands thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P391-Collect spillage.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetonitrile, copper(II) sulphate pentahydrate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.