Learn More
Thermo Scientific™ RapID™ QC Set
Obtain faster recovery, enhanced viability, and direct-streak-to-plate capabilities with QC organisms manufactured using an unmatched proprietary gel preservation process.
Brand: Thermo Scientific™ R4653056
368.13 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 65
Additional Details : Weight : 0.01000kg Transport : UN number : 3373 Chem class : 6.2
Description
Ensure high-performance products, reproducible clinical test results and consistent microbiological evaluation of pharmaceuticals and food with Thermo Scientific™ Culti-Loops™ quality control organisms. Thermo Scientific™ RapID™ QC Sets include a ready-to-use set of QC organisms in Culti-Loops format which are ready-to-use, disposable bacteriological loops containing stabilized, viable microorganisms in a unique gel matrix.
Trust Thermo Scientific quality control organisms for quality results you can depend on, backed by over 30 years of expertise in the manufacturing and packaging of controlled microorganisms.- Decrease time and unnecessary costs of laboratory resources with a direct-streak inoculating loop that requires no rehydration.
- Obtain enhanced viability and recovery with proprietary gel-preserved organisms.
- Consolidate ordering time and shipping costs by turning to one supplier for high-quality culture media and quality control organisms.
- Reach complete traceability with ATCC™ License Derivative strains.
- Achieve accurate and convenient documentation with peel-off, transferable labels.
- Reduce the risk of contamination and infection with zero organism handling.
- More than 600 quality control organisms, including convenient sets for commonly used identification systems
- ISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered
- ATCC™ Licensed Derivative Program member
Specifications
Enterobacter aerogenes, Escherichia coli, Proteus vulgaris, Pseudomonas aeruginosa | |
ISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered | |
RapID™ ONE QC Set/4 | |
Enterobacter aerogenes, Escherichia coli, Proteus vulgaris, Pseudomonas aeruginosa | |
Qualitative QC | |
5 Loops/Pk. |
6380TM* | |
2°C to 8°C | |
Culti-Loops | |
Organism Sets | |
5 Loops/Pack |