missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ RapidFinder™ Beef ID Kit
The RapidFinder™ Beef ID Kit enables the simple and reliable detection of beef in food and feed samples.
Brand: Thermo Scientific™ A24391
497.53 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
- Enables the simple and reliable detection of beef in food and feed samples
- Method works by detection of bovine, or Bos taurus, DNA by real-time PCR
- Unlike ELISA-based methods, when used with the GMO Extraction Kit (4466336) for sample preparation, the RapidFinder™ Beef ID Kit detects both raw and processed beef and provides sensitivity down to 0.01% bovine DNA
- The included positive control, which comprises 0.1% bovine DNA, can be used as a reference to set the threshold of detection of the assay
- Simple-real-time PCR based assay, no electrophoresis needed
- The assay is compatible with Applied Biosystems™ StepOne™, StepOnePlus™, 7500, 7500 Fast, and 7300 real-time PCR systems
Includes
- Beef Master Mix (store at -20°C)
- General Master Mix (store at 4°C or -20°C)
- Positive Control (store at -20°C)
Specifications
Contains 1 tube each of: Beef Master Mix (store at -20°C) General Master Mix (store at 4°C or -20°C) Positive Control (store at -20°C) |
|
Tube | |
qPCR | |
7500 Fast System, 7500 System, StepOne™ System, StepOnePlus™ System | |
Beef Identification Kit |
Primer-probe | |
Low | |
Standard | |
RapidFinder™ | |
Food & Environmental Samples |