missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ RapidFinder™ Pork ID Kit
The RapidFinder™ Pork ID Kit enables the simple and reliable detection of pork in food and feed samples.
Brand: Thermo Scientific™ A24392
494.41 GBP valid until 2024-08-31
Use promo code "21655" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
- Enables the simple and reliable detection of pork in food and feed samples
- The method works by detection of porcine, or Sus scrofa, DNA by real-time PCR
- Unlike ELISA based methods, when used with the GMO Extraction Kit (4466336) for sample preparation, the RapidFinder™ Pork ID Kit detects both raw and processed pork and provides sensitivity down to 0.01% porcine DNA
- The included positive control, which comprises 0.1% porcine DNA, can be used as a reference to set the threshold of detection of the assay
- Simple-real-time PCR based assay, no electrophoresis needed
- The assay is compatible with Applied Biosystems™ StepOne™, StepOnePlus™, 7500, 7500 Fast, and 7300 real-time PCR systems
Includes
- Pork Master Mix (store at -20°C)
- General Master Mix (store at 4°C or -20°C)
- Positive Control (store at -20°C)
Specifications
Contains 1 tube each of: Pork Master Mix (store at -20°C) General Master Mix (store at 4°C or -20°C) Positive Control (store at -20°C) |
|
Tube | |
qPCR | |
7500 Fast System, 7500 System, StepOne™ System, StepOnePlus™ System | |
Pork Identification Kit | |
Food & Environmental Samples |
Primer-probe | |
Low | |
Standard | |
RapidFinder™ | |
48 Reactions |