missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASSF9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528437
This item is not returnable.
View return policy
Description
Recommended positive controls: PAMCI tansfected 293T cell. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene localizes to perinuclear endosomes. This protein associates with peptidylglycine alpha-amidating monooxygenase, and may be involved with the trafficking of this enzyme through secretory or endosomal pathways.Specifications
RASSF9 | |
Polyclonal | |
Unconjugated | |
RASSF9 | |
AW322107; PAM COOH-terminal interactor protein 1; Pamci; PCIP1; P-cip1; Peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor; peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor protein-1; Ras association (RalGDS/AF-6) domain family (N-terminal) member 9; Ras association domain family member 9; ras association domain-containing protein 9; RASSF9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9182 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.4 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O75901 | |
RASSF9 | |
Synthetic peptide corresponding to a region within amino acids 371 and 435 of Human PAMCI | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |