missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat anti-Mouse Kappa Light Chain Secondary Antibody, HRP
Rat Monoclonal Secondary Antibody
Brand: Invitrogen™ MA134732
This item is not returnable.
View return policy
Description
Does not cross-react with other mouse immunoglobulin classes or subclasses. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Mouse Kappa Light Chain | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal | |
Rat | |
Affinity chromatography | |
RUO | |
Mouse | |
Secondary Antibody | |
IgG2a |
Western Blot, ELISA, Immunohistochemistry (Frozen) | |
LO-MK-1 | |
HRP | |
Iglc | |
Purified mouse IgG1 kappa. | |
250 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
AC4647774B | Certificate of Analysis | 08/03/2025 | |
AB4644486D | Certificate of Analysis | 28/02/2025 | |
AB4626088E | Certificate of Analysis | 12/02/2025 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Rat anti-Mouse Kappa Light Chain Secondary Antibody, HRP > 250 μg; HRP
Spot an opportunity for improvement?Share a Content Correction