missing translation for 'onlineSavingsMsg'
Learn More

Rat anti-Mouse IgG1, Clone: A85-1, Secondary Antibody, BD

Brand:  BD Biosciences 553440

Product Code. 15807228

  • £279.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The A85-1 antibody reacts specifically with mouse IgG1 of Igh-Ca and Igh-Cb haplotypes. It does not react with other Ig isotypes. Detection of surface immunoglobulin on B lymphoma cells has been demonstrated with A85-1 mAb. A suspension of pooled mouse Ig

Host Species: Rat
Clone: A85-1
Isotype: IgG1, κ
Species Reactivity: Mouse

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rat anti-Mouse IgG1
A85-1
Rat
0.5 mg
Immunology
Mouse
Monoclonal
0.5mg/mL
Affinity Purified
RUO
Secondary
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers