Learn More
Invitrogen™ Rat anti-Mouse IgG1 (Heavy chain) Secondary Antibody, FITC
Rat Monoclonal Secondary Antibody
Brand:  Invitrogen™ MA516785
Product Code. 15276029
  • £552.00 / 500µg
Explore more special offers
Description
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse IgG1 Heavy Chain antibody, clone LO-MG1-2 recognizes murine IgG1, and does not bind other mouse immunoglobulin classes or subclasses.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse IgG1 (Heavy chain)
Monoclonal
1 mg/mL
PBS with 50% glycerol and <0.1% sodium azide
Purified murine IgG1 from BALB/c mice.
500 μg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Liquid
Flow Cytometry, Immunocytochemistry
LO-MG1-2
FITC
Rat
Affinity chromatography
RUO
Mouse
Secondary Antibody
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Rat anti-Mouse IgG1 (Heavy chain) Secondary Antibody, FITC > 500 μg; FITC