missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat anti-Mouse IgG1 (Heavy chain) Secondary Antibody, HRP
Rat Monoclonal Secondary Antibody
Brand: Invitrogen™ MA516786
This item is not returnable.
View return policy
Description
This product may be used in a direct ELISA or as a detection reagent in a sandwich ELISA together with MA5-16782 as the capture reagent. Rat anti Mouse IgG1 Heavy Chain antibody, clone LO-MG1-2 recognizes murine IgG1, and does not bind other mouse immunoglobulin classes or subclasses.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Mouse IgG1 (Heavy chain) | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal | |
Monoclonal mouse IgG1. | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
LO-MG1-2 | |
HRP | |
Rat | |
Affinity chromatography | |
RUO | |
Mouse | |
Secondary Antibody | |
IgG1 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
162330 | Certificate of Analysis | 27/03/2025 | |
AC4660493B | Certificate of Analysis | 20/03/2025 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Rat anti-Mouse IgG1 (Heavy chain) Secondary Antibody, HRP > 500 μg; HRP
Spot an opportunity for improvement?Share a Content Correction