Learn More
Rat anti-Mouse IgG1, Secondary Antibody, BD
Brand: BD Biosciences 562580
Additional Details : Weight : 0.00820kg
Description
The A85-1 antibody reacts specifically with mouse IgG1 of Igh-Ca and Igh-Cb haplotypes. It does not react with other Ig isotypes. Detection of surface immunoglobulin on B lymphoma cells has been demonstrated with the A85-1 monoclonal antibody. A suspension of pooled mouse IgG1 was used as the source of immunogen.
Host Species: Rat
Clone: A85-1
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Pooled Mouse IgG1
Flow Cytometry
Specifications
Mouse IgG1 | |
A85-1 | |
Brilliant Violet 421 | |
Affinity Purified | |
RUO | |
Mouse |
Monoclonal | |
0.2mg/mL | |
Rat | |
50 μg | |
Secondary | |
IgG1 κ |