Learn More
Invitrogen™ Rat anti-Mouse IgG2a (Heavy chain) Secondary Antibody, HRP
Rat Monoclonal Secondary Antibody
Brand: Invitrogen™ MA516790
Description
Rat anti Mouse IgG2a:Biotin, clone LO-MG2a-7 recognizes the gamma 2a heavy chain of mouse immunoglobulin and does not cross-react with other murine immunoglobulin classes or subclasses.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Mouse IgG2a (Heavy chain) | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal | |
Purified IgG from BALB/c mice. | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
LO-MG2a-7 | |
HRP | |
Rat | |
Affinity chromatography | |
RUO | |
Mouse | |
Secondary Antibody | |
IgG1 |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
AC4660494A | Certificate of Analysis | 25/03/2025 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.