Learn More
Rat anti-Mouse IgM, Secondary Antibody, BD
Brand: BD Biosciences 562595
Additional Details : Weight : 0.00820kg
Description
The R6-60.2 antibody monoclonal antibody specifically binds to mouse Immunoglobulin M (IgM) of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes. The R6-60.2 antibody has not been shown to stimulate B-cell proliferation.
Host Species: Rat
Clone: R6-60.2
Isotype: IgG2a, κ
Species Reactivity: Mouse
Immunogen: Pooled Mouse Ig
Flow Cytometry
Specifications
Mouse IgM | |
R6-60.2 | |
Brilliant Violet 421 | |
Affinity Purified | |
RUO | |
Mouse |
Monoclonal | |
0.2mg/mL | |
Rat | |
50 μg | |
Secondary | |
IgG2a κ |