missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Rat CCL5 (RANTES) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 400-13-20UG
142.20 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
RANTES is a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. It binds to CCR1, CCR3, CCR4 and CCR5 and is one of the major HIV suppressive factors produced by CD8+ T cells. Recombinant RANTES protein induces a dose dependent inhibition of different strains of HIV 1, HIV 2, and simian immunodeficiency virus (SIV). It is T cell and macrophage specific, induced by mitogens, and belongs to the intercrine beta (chemokine CC) family.Specifications
P50231 | |
protein with no preservative | |
7.9 | |
20 μg | |
-20°C | |
<1 EU/μg | |
CCL5 | |
Unconjugated | |
SPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMS | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
81780 | |
Rat CCL5 (RANTES) | |
E. coli | |
RUO | |
beta-chemokine RANTES; CC chemokine; C-C motif chemokine 5; C-C motif chemokine ligand 5; CCL5; chemokine (C-C motif) ligand 5; D17S136E; EoCP; eosinophil chemotactic cytokine; MGC17164; MuRantes; OTTHUMP00000197106; RANTES; RANTES protein; RANTES(3-68); RANTES(4-68); regulated upon activation normal T-cell expressed and secreted; regulated upon activation, normally T-expressed, and presumably secreted; Scya5; SIS d; SISd; SIS-delta; SISd; small inducible cytokine A5; small inducible cytokine subfamily A (Cys-Cys), member 5; small-inducible cytokine A5; T cell-specific protein P228; T-cell specific protein p288; T-cell-specific protein RANTES; TCP228; Unknown (protein for MGC:127014) | |
Rat | |
Recombinant | |
Lyophilized |