Learn More
R&D Systems™ Rat Cortical Stem Cells (3 x 10e6 cells/vial)
For cortical stem cell expansion and differentiation into neurons, astrocytes and oligodendrocytes
Brand: R&D Systems™ NSC001
Additional Details : Weight : 0.00970kg
Description
- Have been fully characterized to reduce experimental variation
- Can be expanded as monolayers or neurospheres
- Are free of mycoplasma and microbial contamination
Specifications
3 x 10e6 cells/vial |