missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IgG2b kappa Isotype Control (eB149/10H5), Biotin, eBioscience™, Invitrogen™
Rat Isotype Control
£47.64 - £220.00
Specifications
Antigen | Rat IgG2b kappa |
---|---|
Clone | eB149/10H5 |
Conjugate | Biotin |
Form | Liquid |
Host Species | Rat |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15299789
|
Invitrogen
13-4031-81 |
50 μg |
£47.64
50µg |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
15209799
|
Invitrogen
13-4031-82 |
100 μg |
£89.50
100µg |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
15219799
|
Invitrogen
13-4031-85 |
500 μg |
£220.00
500µg |
Estimated Shipment: 01-11-2024
Log in to see stock. |
|||||
Beschreibung
Description: The rat IgG2b, kappa monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: Biotin Rat IgG2b Isotype Control has been reported for use in immunohistochemistry, immunocytochemistry and flow cytometric analysis. Applications Tested: Biotin Rat IgG2b Isotype Control has been tested by flow cytometric analysis of mouse splenocyte suspensions. Use at the same concentration as the experimental antibody. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2b is a protein-coding gene that encodes for the immunoglobulin G2b (IgG2b) subclass of rat antibodies. IgG2b is a major subclass of rat immunoglobulins that plays a crucial role in the immune response by neutralizing pathogens and promoting opsonization. Rat IgG2b is commonly used in research applications, such as in immunohistochemistry, flow cytometry, and Western blotting.Spezifikation
Rat IgG2b kappa | |
Biotin | |
Rat | |
IgG2b κ | |
Affinity chromatography |
eB149/10H5 | |
Liquid | |
RUO | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.