missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat IL-9 ELISA Kit
Rat IL-9 quantitates rat IL-9 in serum, plasma, supernatant.
£530.00 - £2320.00
Specifications
Accession Number | cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; Il9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor p40 |
---|---|
Assay Range | 0.45 to 100 ng/mL |
Assay Sensitivity | 0.45 ng/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17175869
|
Invitrogen™
ERA34RBX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
17195043
|
Invitrogen™
ERA34RB |
96 Tests |
£530.00
Pack of 96 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
Description
Rat IL-9 quantitates rat IL-9 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat IL-9.
Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.Specifications
cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; Il9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor p40 | |
0.45 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.45 to 100 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Il9 | |
<10% | |
HRP | |
Supernatant, 50 μL; Plasma, 50 μL; Serum, 50 μL | |
IL-9 | |
4 hr 45 min |