missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat IL-9 ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ ERA34RBX10
Additional Details : Weight : 2.50000kg
Description
Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.Specifications
Q00993 | |
0.45 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Il9 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
0.45 to 100 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
116558 | |
<12% | |
Pre-coated 96 well plate|Standard|Assay Diluent concentrate|Biotinylated Detection Antibody|SAV-HRP|Wash Buffer|Chromogen|Stop Solution|Adhesive Plate Covers | |
10 x 96 Tests | |
Supernatant, 50 μL|Plasma, 50 μL|Serum, 50 μL | |
IL-9 | |
4 hr 45 min |