Learn More
Invitrogen™ Rat MINT3 (aa 1-19) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP135
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Rat Mint3 (residues 1-19) Synthetic Peptide, PEP-135, from Invitrogen.
PEP-135 is a 19 amino acid synthetic peptide whose sequence corresponds to amino acid residues 1-19 from rat Mint3 protein. The sequence of this peptide is (amino to carboxy terminus): M (1)-E-F-L-P-E-P-Q-H-P-P-G-P-P-T-M-D-L- E (19).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and rat Mint3 protein, catalog # PA1-072. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
83611 | |
purified | |
Rat MINT3 (aa 1-19) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Rat | |
O70248 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Apba3 | |
No preservative | |
50 μg | |
M(1) E F L P E P Q H P P G P P T M D L E(19) |